SARS nucleocaspid
Supplier: Biorbyt
Certificates
About this item
Specifications
- Storage conditions:Five years frozen. One month at 4 °C.
- Protein/peptide name:SARS Nucleocaspid
- Purity:SARS Core protein is >95% pure as determined by 10% PAGE (coomassie staining).
- Formulation:50mM Tris-HCl, 60mM NaCl and 50% glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together