SARS envelope
Supplier: Biorbyt
Certificates
About this item
Specifications
- Storage conditions:Five years frozen. One month at 4 °C.
- Protein/peptide name:SARS Envelope
- Purity:SARS Envelope protein is >95% pure as determined by 10% PAGE(coomassie staining). SARS Envelope protein was purified by proprietary chromatographic technique.
- Formulation:50mM Tris-HCl, 60mM NaCl, and 50% glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together