HIV-1 p17/24/120 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Storage conditions:HIV-1 p17,p24, gp120 protein is shipped at ambient temperature. Upon arrival, Store at −20 °C Stable for Five years frozen. One month in solution at room temperature.
- Protein/peptide name:HIV-1 p17/24/120
- Purity:Greater than 95.0% as determined by HPLC analysis and SDS-PAGE.
- Formulation:8M urea, 25mM Tris-HCl pH 8.0, 5mM beta mercaptoethanol.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together