HTLV-1 p24c
Supplier: Biorbyt
Certificates
About this item
Specifications
- Storage conditions:Five years frozen. One month in solution at room temperature.
- Protein/peptide name:HTLV-1 p24c
- Purity:HTLV-1 p24 protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. HTLV-1 p24 was purified by proprietary chromatographic technique.
- Formulation:50mM NaPO4 pH 6.0, containing 1mM DTT 1mM EDTA.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together