Dengue envelope-2 45 kDa (from E. coli)
: Biorbyt
- Source:E. coli
- Storage conditions:Five years frozen. One month in solution at room temperature.
- Protein/peptide name:Dengue Envelope-2 45kDa
- Purity:Protein is >95% pure as determined by 10% PAGE (coomassie staining).
- Formulation:Phosphate buffered saline, pH-7.4.
- Tested applications:SDS-PAGE , HPLC , FuncS
Frequently Bought Together