Dengue virus subtype envelope 15 kDa, (Domain III)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Storage conditions:Five years frozen. One month in solution at room temperature.
- Protein/peptide name:Dengue Virus Subtype Envelope 15kDa, (Domain III)
- Purity:Protein is >95% pure as determined by 10% PAGE (coomassie staining).
- Sequence:VMCTGSFKLE KEVAETQHGT VLVQVKYEGT DAPCKIPFST QDEKGVTQNG RLITANPIVT DKEKPVNIEA EPP FGESYI VVGAGEKALK LSWFKKGSV.
- Formulation:50mM sodium chloride-phosphorous buffer, pH-7.4.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together