Chlamydia W3-6
: Biorbyt
- Storage conditions:Five years frozen. One month in solution at room temperature.
- Protein/peptide name:Chlamydia W3-6
- Purity:Chlamydia W3-W6 protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. Purifications Method Chlamydia W3-W6 protein was purified by proprietary chromatographic technique.
- Formulation:10mM Tris-HCl, pH 6.5, 100mM Sodium Phosphate and 8M urea.
- Tested applications:SDS-PAGE , HPLC , FuncS
Frequently Bought Together