Human recombinant ISG15 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:ISG15 although stable 4 °C for weeks, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Protein synonyms:G1P2
- Protein/peptide name:ISG15
- Purity:Greater than 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Sequence:MADQLTEEQI AEFKEAFSLF DKDGDGTITT KELGTVMRSL GQNPTEAELQ DMINEVDADG NGTIDFPEFL TMMARKMKDT DSEEEIREAF RVFDKDGNGY ISAAELRHVM TNLGEKLTDE EVDEMIREAD IDGDGQVNYE EFVQMMTAKG SMGWDLTVKM LAGNEFQVSL SSSMSVSELK AQITQKIGVH AFQQRLAVHP SGVALQDRVP LASQGLGPGS TVLLVVDKCD EPLNILVRNN KGRSSTYEVR LTQTVAHLKQ QVSGLEGVQD DLFWLTFEGK PLEDQLPLGE YGLKPLSTVF MNLRLRGG.
- Formulation:The ISG15 solution contains 20 mM Tris-HCl pH-8, 100mM NaCl and 1mM DTT.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together