Order Entry
Austria
ContactUsLinkComponent
Human recombinant TIM3 (from cells)
Human recombinant TIM3 (from cells)
Catalog # BIRBORB180419-50
Supplier:  Biorbyt
CAS Number:  
Human recombinant TIM3 (from cells)
Catalog # BIRBORB180419-50
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    T-Cell Membrane
  • UniProtKB:
    Q8TDQ0 (Human)
  • Protein/peptide name:
    TIM3
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    SEVEYRAEVGQNAYLPCFYTPAAPGNLVPVCWGKGACPVFECGNVVLRTDERDVNYWTSRYWLNG DFRKGDVSLTIENVTLADSGIYCCRIQIPGIMNDEKFNLKLVIKPAKVTPAPTLQRDFTAAFPRM LTTRGHGPAETQTLGSLPDINLTQISTLANELRDSRLANDLRDSGATIRVDDIEGRMDEPKSCDK THTCPPCPAPELLGGPSVFLFPPKPKDTLMISRTPEVTCVVVDVSHEDPEVKFNWYVDGVEVHNA KTKPREEQYNSTYRVVSVLTVLHQDWLNGKEYKCKVSNKALPAPIEKTISKAKGQPREPQVYTLP PSREEMTKNQVSLTCLVKGFYPSDIAVEWESNGQPENNYKTTPPVLDSDGSFFLYSKLTVDKSRW QQGNVFSCSVMHEALHNHYTQKSLSLSPGKHHHHHH
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4.
  • Tested applications:
    SDS-PAGE , HPLC

Specifications

About this item