Mouse recombinant FGF18 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:Murine Fibroblast Growth Factor-18
- Protein/peptide name:FGF18
- Purity:> 95 % by SDS-PAGE and HPLC analyses
- Molecular weight:21.0 kDa
- Sequence:EENVDFRIHV ENQTRARDDV SRKQLRLYQL YSRTSGKHIQ VLGRRISARG EDGDKYAQLLVETDTFGSQV RIKGKETEFY LCMNRKGKLV GKPDGTSKEC VFIEKVLENN YTALMSAKYSGWYVGFTKKG RPRKGPKTRE NQQDVHFMKR YPKGQAELQK PFKYTTVTKRSRRIRPTHPG
- Formulation:Lyophilized from a 0.2 m filtered concentrated solution in PBS, pH 7.4, 500 mM NaCl
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together