Order Entry
Austria
ContactUsLinkComponent
Mouse recombinant TECK (from E. coli)
Mouse recombinant TECK (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse recombinant TECK (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    Murine Thymus Expressed Chemokine
  • Protein/peptide name:
    TECK
  • Purity:
    > 95% by SDS-PAGE and HPLC analyses.
  • Molecular weight:
    14.1 kDa
  • Sequence:
    QGAFEDCCLG YQHRIKWNVL RHARNYHQQE VSGSCNLRAV RFYFRQKVVCGNPEDMNVKR AIRILTARKR LVHWKSASDS QTERKKSNHM KSKVENPNSTSVRSATLGHP RMVMMPRKTN N
  • Formulation:
    Lyophilized from a 0.2 um filtered concentrated solution in 20mM PB, pH 7.4, 150mM NaCl
  • Tested applications:
    SDS-PAGE , HPLC

Specifications

Related Information

Frequently Bought Together