Rat recombinant IL1 alpha (from E.coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:Hematopoietin 1
- Protein/peptide name:IL1 alpha
- Purity:> 97 % by SDS-PAGE and HPLC analyses
- Molecular weight:17.8 kDa
- Sequence:SAPHSFQNNL RYKLIRIVKQ EFIMNDSLNQ NIYVDMDRIH LKAASLNDLQLEVKFDMYAY SSGGDDSKYP VTLKVSNTQL FVSAQGEDKP VLLKEIPETPKLITGSETDL IFFWEKINSK NYFTSAAFPE LLIATKEQSQ VHLARGLPSMIDFQIS
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in 20 mM Tris, 150 mM NaCl, pH 8.5
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together