0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Proteins and Peptides
PAPP-A
PAPP-A
BIRBORB79680-1
:
Biorbyt
:
PAPP-A
BIRBORB79680-1
:
Biorbyt
:
Pk:
1 mg
Storage conditions:
Store at -2 to -8 °C.
Protein/peptide name:
PAPP-A
Purity:
Greater than 95% pure SDS-PAGE. Protein G Chromatography