Order Entry
Austria
ContactUsLinkComponent
 
GPBB
  BIRBORB79666-1
 :  Biorbyt
 :  
GPBB
  BIRBORB79666-1
 :  Biorbyt
 :  

 

  • Pk:
    1 mg
  • Storage conditions:
    Store at -2 to -8 °C.
  • Protein/peptide name:
    GPBB
  • Purity:
    Greater than 95% pure SDS-PAGE. Protein G Chromatography