Human recombinant HGb (from E. coli)
Lieferant: Biorbyt
Zertifikate
Über diesen Artikel
Spezifikationen
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Cytoglobin
- UniProtKB:Q8WWM9 (Human)
- Protein/peptide name:HGb
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:MEKVPGEMEIERRERSEELSEAERKAVQAMWARLYASCEDVGVAILVRFFVNFPSAKQYFSQFKH MEDPLEMERSPQLRKHACRVMGALNTVVENLHDPDKVSSVLALVGKAHALKHKVEPVYFKILSGV ILEVVAEEFASDFPPETQRAWAKLRGLIYSHVTAAYKEVGWVQQVPNATTPPATLPSSGPLEHHH HHH
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 10% Glycerol, pH 8.0 .
- Tested applications:SDS-PAGE
Spezifikationen
Frequently Bought Together