Human recombinant XPNPEP1 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:XaaPro Aminopeptidase 1
- UniProtKB:Q9NQW7 (Human)
- Protein/peptide name:XPNPEP1
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:PPKVTSELLRQLRQAMRNSEYVTEPIQAYIIPSGDAHQSEYIAPCDCRRAFVSGFDGSAGTAIIT EEHAAMWTDGRYFLQAAKQMDSNWTLMKMGLKDTPTQEDWLVSVLPEGSRVGVDPLIIPTDYWKK MAKVLRSAGHHLIPVKENLVDKIWTDRPERPCKPLLTLGLDYTGISWKDKVADLRLKMAERNVMW FVVTALDEIAWLFNLRGSDVEHNPVFFSYAIIGLETIMLFIDGDRIDAPSVKEHLLLDLGLEAEY RIQVHPYKSILSELKALCADLSPREKVWVSDKASYAVSETIPKDHRCCMPYTPICIAKAVKNSAE SEGMRRAHIKDAVALCELFNWLEKEVPKGGVTEISAADKAEEFRRQQADFVDLSFPTISSTGPNG AIIHYAPVPETNRTLSLDEVYLIDSGAQYKDGTTDVTRTMHFGTPTAYEKECFTYVLKGHIAVSA AVFPTGTKGHLLDSFARSALWDSGLDYLHGTGHGVGSFLNVHEGPCGISYKTFSDEPLEAGMIVT DEPGYYEDGAFGIRIENVVLVVPVKTKYNFNNRGSLTFEPLTLVPIQTKMIDVDSLTDKECDWLN NYHLTCRDVIGKELQKQGRQEALEWLIRETQPISKQHHHHHH
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 10% Glycerol, pH 8.0.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together