Order Entry
Austria
ContactUsLinkComponent
Human recombinant IL2 (fromE. coli)
Human recombinant IL2 (fromE. coli)
Supplier:  Biorbyt
Certificates
Human recombinant IL2 (fromE. coli)
Supplier:  Biorbyt

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Biological activity:
    ED50 is less than 0.1 ng/ml as determined by the dosedependent stimulation of murine CTLL2 cells.
    Specific Activity of 1.0 x 107 IU/ mg.
  • Protein synonyms:
    Interleukin2
  • UniProtKB:
    P60568 (Human)
  • Protein/peptide name:
    IL2
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MPTSSSTKKTQLQLEHLLLDLQMILNGINNYKNPKLTRMLTFKFYMPKKATELKHLQCLEEELKP LEEVLNLAQSKNFHLRPRDLISNINVIVLELKGSETTFMCEYADETATIVEFLNRWITFCQSIIS TLT
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM HAc-NaAc, 150mM NaCl, pH 4.0.
  • Tested applications:
    SDS-PAGE , FuncS

Specifications

Related Information

Frequently Bought Together