Human recombinant BID (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Apoptic death agonist
- UniProtKB:P55957 (Human)
- Protein/peptide name:BID
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:MDCEVNNGSSLRDECITNLLVFGFLQSCSDNSFRRELDALGHELPVLAPQWEGYDELQTDGNRSS HSRLGRIEADSESQEDIIRNIARHLAQVGDSMDRSIPPGLVNGLALQLRNTSRSEEDRNRDLATA LEQLLQAYPRDMEKEKTMLVLALLLAKKVASHTPSLLRDVFHTTVNFINQNLRTYVRSLARNGMD
- Formulation:Supplied as a 0.2 um filtered solution of 20mM PB, 100mM KCl, pH 7.4.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together