Order Entry
Austria
ContactUsLinkComponent
Human recombinant APE (from E. coli)
Human recombinant APE (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant APE (from E. coli)
Supplier:  Biorbyt

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    DNA(Apurinic or Apyrimidinic Site) Lyase
  • UniProtKB:
    P27695 (Human)
  • Protein/peptide name:
    APE
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MGPKRGKKGAVAEDGDELRTEPEAKKSKTAAKKNDKEAAGEGPALYEDPPDQKTSPSGKPATLKI CSWNVDGLRAWIKKKGLDWVKEEAPDILCLQETKCSENKLPAELQELPGLSHQYWSAPSDKEGYS GVGLLSRQCPLKVSYGIGEEEHDQEGRVIVAEFDSFVLVTAYVPNAGRGLVRLEYRQRWDEAFRK FLKGLASRKPLVLCGDLNVAHEEIDLRNPKGNKKNAGFTPQERQGFGELLQAVPLADSFRHLYPN TPYAYTFWTYMMNARSKNVGWRLDYFLLSHSLLPALCDSKIRSKALGSDHCPITLYLAL
  • Formulation:
    Supplied as a 0.2 um filtered solution of 10mM HEPES, 100mM KCl, 50% Glycerol, pH 7.4.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together