G-CSF
BIRBORB97766-10
: Biorbyt
:
- Pk:10 µG
- Storage conditions:Store at−20 °C.
- Biological activity:ED50: Less than 0.1 ng/ml corresponding to a specific activity of 1x108 IU/mg, calculated by the dosedependantproliferation of murine NFS-60 indicator cells measuredby 3H-thymidine uptake
- UniProtKB:P09919
- Protein/peptide name:G-CSF
- Purity:95% pure by SDS-PAGE and RP-HPLC
- Solubility:It is recommended to reconstitute the lyophilized G-CSFin sterile bidest water not less than 100 g/ml, which canthen be further diluted to other aqueous solutions. Forlong term storage it is recommended to add a carrierprotein (0.1% HSA or BSA).
- Molecular weight:19 kDa
- Formulation:Lyophilized. G-CSF was lyophilized after dialysis against10 mM sodium acetate pH 4.0.