Vitamin D binding protein (Gc-globulin) (from plasma)
Catalog # BIRBORB82153-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Source:Plasma
- Species:
- Environmentally Preferable:
- Protein/peptide name:Vitamin D Binding Protein (Gc-Globulin)
- Purity:Purified.
- Formulation:Lyophilised.
- Tested applications:SDS-PAGE