Human myelin basic protein (from brain)
Catalog # BIRBORB82267-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Source:Brain
- Species:Human
- Protein synonyms:Myelin Basic
- Protein/peptide name:Myelin Basic Protein
- Purity:>90%
- Formulation:Lyophilised. Reconstitute with 10 mM HCl.