Order Entry
Austria
ContactUsLinkComponent
Human recombinant Apolipoprotein E (from Plasma)
Human recombinant Apolipoprotein E (from Plasma)
Catalog # BIRBORB82120-50
Supplier:  Biorbyt
CAS Number:  
Human recombinant Apolipoprotein E (from Plasma)
Catalog # BIRBORB82120-50
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Plasma
  • Species:
    Human
  • Protein synonyms:
    AD2
  • Protein/peptide name:
    Apolipoprotein E
  • Purity:
    Purified.
  • Formulation:
    Not lyophilised.

Specifications

About this item