Human apolipoprotein AI (from Plasma)
Catalog # BIRBORB82152-0.5
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:0,5 mg
- Source:Plasma
- Species:Human
- Protein synonyms:APOA1
- Protein/peptide name:Apolipoprotein AI
- Purity:Purified.
- Formulation:Not lyophilised.
- Tested applications:SDS-PAGE