Recombinant herpes simplex virus-1 (HSV-1) glycoprotein D (from E. coli)
Catalog # BIRBORB82476-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:E. coli
- Protein/peptide name:Herpes Simplex Virus-1 (HSV-1) Glycoprotein D
- Purity:Purified.
- Formulation:Not lyophilised.
- Tested applications:ELISA , WB