Order Entry
Austria
ContactUsLinkComponent
Influenza A H5N1 (Avian) Nonstructural protein 1 (NS1)
Influenza A H5N1 (Avian) Nonstructural protein 1 (NS1)
Catalog # BIRBORB82547-50
Supplier:  Biorbyt
CAS Number:  
Influenza A H5N1 (Avian) Nonstructural protein 1 (NS1)
Catalog # BIRBORB82547-50
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein synonyms:
    Influenza A H5N1 (Avian) Nonstructural 1 (NS1)
  • Protein/peptide name:
    Influenza A H5N1 (Avian) Nonstructural Protein 1 (NS1)
  • Purity:
    Purified.
  • Formulation:
    Preservative: 0.02% Sodium azide
  • Tested applications:
    ELISA

Specifications

About this item