Order Entry
Austria
ContactUsLinkComponent
Rat IL6 (from E. coli)
Rat IL6 (from E. coli)
Supplier:  Biorbyt
Certificates
Rat IL6 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Rat
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The ED50= 0.03-0.1 ng/mL. The biological activity is determined by measuring the dose-dependant proliferation of IL-6 dapendent B9 cells. concentration range of 0.1 to 10.0 ng/mL is effective for most in vitro applications.
  • Protein synonyms:
    Interleukin BSF 2
  • Protein/peptide name:
    IL6
  • Purity:
    > 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Interleukin-6 in 100mM acetic acid to 1.0 mg/mL and incubated for 30 minutes at room temperature to regain full activity, which can then be further diluted to other aqueous solutions.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be Met-Phe-Pro-Thr-Ser.
  • Formulation:
    The protein was lyophilized without any additives.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together