Order Entry
Austria
ContactUsLinkComponent
Rat FGF basic (from E. coli)
Rat FGF basic (from E. coli)
Supplier:  Biorbyt
Certificates
Rat FGF basic (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Rat
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The ED50 range as determined by the dose-dependant proliferation of BALB/3T3 cells was found to be less than 0.2 ng/ml, corresponding to specific activity of 5,000,000 IU/mg.
  • Protein synonyms:
    HBGH-2
  • Protein/peptide name:
    FGF basic
  • Purity:
    > 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Rat bFGF in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    PALPEDGGGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHV KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTEE CFFFERLESN NYNTYRSRKY SSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS.
  • Formulation:
    FGF-b was lyophilized from mg/ml solution after extensive dialysis against 20 mM phosphate buffer, pH 7.4 and 130mM NaCl.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together