Human recombinant SNCG (from E. coli)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA). Avoid multiple freeze-thaw cycles.
- Protein synonyms:Gammalpha Synuclein
- Protein/peptide name:SNCG
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Sequence:mdvfkkgfsi akegvvgave ktkqgvteaa ektkegvmyv gaktkenvvqsvtsvaektk eqanavseav vssvntvatk tveeaeniav tsgvvrkedl rpsapqqegv askekeevae eaqsggd.
- Formulation:The protein (1mg/ml) contains 20 mM Tris-HCl buffer (pH 7.5), 0.1M NaCl and 1mM MgCl2.
- Tested applications:SDS-PAGE
Frequently Bought Together