Order Entry
Austria
ContactUsLinkComponent
Testosterone ELISA kit
Testosterone ELISA kit
Catalog # ABNOKA0309
Supplier:  Abnova
CAS Number:  
undefined
Testosterone ELISA kit
Catalog # ABNOKA0309
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    Testosterone
  • Description:
    Testosterone ELISA Kit, 96 wells

Specifications

About this item

The Testosterone ELISA Kit is a colorimetric, competitive immunoassay for the quantitative determination of testosterone in biological fluids. These are suitable for the quantitative determination of testosterone in culture supernatants, plasma, serum, and saliva from any species.

  • Easy to use with convenient storage at 4 °C
  • Calibration range: 7,81 - 2000 pg/ml
  • Sample volume: 100 µl
  • Limit of detection: 5,67 pg/ml
Delivery: Kit contains GxM IgG microtiter plate (96 wells), conjugate (6 ml), antibody (6 ml), assay buffer 3 (30 ml), wash buffer concentrate (27 ml), standard (0,5 ml), steroid displacement reagent (1 ml), pNpp substrate (20 ml), stop solution (5 ml) and plate sealer (1 slice).