Orders LinkContactUsLinkComponent
Cable lockouts, original
Cable lockouts, original
Supplier:   Brady
Cable lockouts, original
Supplier:  Brady
Certificates
Cable lockouts, original
Cable lockouts, original
3 Options Available Below
Product Details & Documents
Brady's original cable lockout is perfect for difficult lockout situations such as large gate valves or oversized devices.
  • Body is made from impact modified, glass-filled nylon; cable is made from galvanised steel
  • One padlock can be applied
  • With 1,8 m galvanised steel cable (3,2 mm outside diameter)
Max. shackle diameter: 8,00 mm
Max. wire/cable diameter: 3,20 mm
W×H: 72×182,70 mm
Product Family Options
Product Information
  • Type
    Colour
    Availability
    Price
  • Cable lockout, with 1,8 mm cable
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,20 €
    70,20 €
    1 * 1 ST
       70,20 €
  • Cable lockout, with 1,8 mm cable
    Red
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,20 €
    70,20 €
    1 * 1 ST
       70,20 €
  • Cable lockout, with 1,8 mm cable
    Yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,20 €
    70,20 €
    1 * 1 ST
       70,20 €