Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Redox/ORP Standards
Redox standards
Redox standards
Redox standards
Catalog # HAMI238322
Supplier:
HAMILTON BONADUZ
CAS Number:
Certificates
Expand image
Redox standards
Catalog # HAMI238322
Supplier:
HAMILTON BONADUZ
CAS Number:
Certificates
Specifications
Pk:
250 ml
Description:
475±5 mV ORP buffer
Specifications
About this item
Product Details
Documentation
Product Details
Documentation