ContactUsLinkComponent
Restriction enzymes, XhoI, FastDigest™, Fermentas
Restriction enzymes, XhoI, FastDigest™, Fermentas
Certificates
undefined
Restriction enzymes, XhoI, FastDigest™, Fermentas
Specifications
  • Compatible buffer:
    FastDigest Green Buffer or FastDigest Buffer (colorless)
  • Overhang type:
    5' overhang
  • Enzyme name:
    XhoI