Order Entry
Austria
ContactUsLinkComponent
Non-interfering Protein Assay™ kit, Calbiochem®
Non-interfering Protein Assay™ kit, Calbiochem®
Catalog # 488250-1
CAS Number:  
Non-interfering Protein Assay™ kit, Calbiochem®
Catalog # 488250-1
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Non-Interfering Protein Assay™ Kit, 500 assays
  • Environmentally Preferable:

Specifications

About this item

Non-Interfering Protein Assay™ Kit is an easy-to-use protein assay that overcomes interference of agents found in protein solutions including detergents, chelating agents, reducing agents, amines, sugars, urea, and others. The Universal Protein Precipitating Agent (UPPA™ Reagent) is used to precipitate and immobilize the protein in the tube while the interfering reagents are removed.

  • Protein concentration is based on the specific binding of copper to the peptide backbone
  • With the increase in protein concentration, concentration of unbound copper ions decreases
  • Colour density is inversely related to the amount of protein present in solution
Delivery: This kit is supplied with UPPA™ reagents I and II, copper solution I, color agents A and B, BSA standard and a user protocol.