Order Entry
Austria
ContactUsLinkComponent
Urease (from Canavalia ensiformis)
Urease (from Canavalia ensiformis)
  666133-10
 :  
Urease (from Canavalia ensiformis)
  666133-10
 :  

 

  • Pk:
    10 KU
  • CAS No.:
    9002-13-5
  • Source:
    Canavalia ensiformis
  • Storage conditions:
    −20 °C
  • Form:
    White lyophilized powder
  • Enzyme name:
    Urease
  • Enzyme activity:
    >50 u/mg material (Nesslers)
  • pH:
    7,0

 

 

Native urease from Canavalia ensiformis. Catalyzes hydrolysis of urea to carbon dioxide and ammonia. Useful in the determination of urea in body fluids.

  • One unit is defined as the amount of enzyme that will release 1 µmol of ammonia from urea per min at 25 °C.