Extendable samplers, TeleSwab
Supplier: Sampling Systems
Jump to:
About this item
The TeleSwab has an extendable handle, which allows to take swabs from high and difficult to reach places.
- Environmental swabbing for use with large items of processing equipment
- Extendable handle to the correct length for easy swabbing
- Adapter allowing the swabs to be angled to the contours of the sampled item
Ordering information: Swabs are not included with these kits.
2 Options Available Below
Product Details & Documents
The TeleSwab has an extendable handle, which allows to take swabs from high and difficult to reach places.
- Environmental swabbing for use with large items of processing equipment
- Extendable handle to the correct length for easy swabbing
- Adapter allowing the swabs to be angled to the contours of the sampled item
Ordering information: Swabs are not included with these kits.
Product Family Options
Product Information
- AvailabilityPrice
- 300-0420Telescopic swabbing tool with extendable pole, swab holder and angle adapterTelescopic swabbing tool with extendable pole, swab holder and angle adapterLog in for availability1 * 1 ST4 390,00 €
- 300-0421Telescopic swabbing tool with extendable pole, swabbing template, template holder and angle adapterTelescopic swabbing tool with extendable pole, swabbing template, template holder and angle adapterLog in for availability1 * 1 ST4 390,00 €