Order Entry
Austria
Orders LinkContactUsLinkComponent
Cable lockouts, all purpose

Cable lockouts, all purpose

Supplier:   Brady
Cable lockouts, all purpose
Supplier:  Brady
Certificates
Cable lockouts, all purpose
Cable lockouts, all purpose

2 Options Available Below

Product Details & Documents

These devices are ideal for locking out multiple control points.

  • Perfect for unusual energy isolating devices that are difficult to lock out
  • Easy to use - simply hold device in one hand and clinch tight with the other
  • Integrated eyelet keeps the cables conveniently wound, preventing the ends from dangling
  • Accepts cables with diameters between 1,59 and 4,77 mm
  • Supports up to four padlocks

 : Supplied with without cable, with vinyl coated metal cable or with non conductive nylon cable.

Product Family Options

Product Information

  • Type
    Colour
    W×D×H
    Availability
    Price
  • All purpose cable lockout with vinyl coated metal cable (2,44 m)
    Red
    82×32,4×152,40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 95,70 €
    95,70 €
    1 * 1 ST
       95,70 €

  • All purpose cable lockout, without cable
    Red
    82×32,4×152,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,00 €
    82,00 €
    1 * 1 ST
       82,00 €

Accessories

Product Information

  • Catalog #
    Colour
    Availability
    Price
  • Non conductive nylon cable, 2,44 m, for use with all purpose cable lockout
    Non conductive nylon cable, 2,44 m, for use with all purpose cable lockout
    BRDY050945
    Red
    Log in for availability
    1 * 1 ST - 22,10 €