Order Entry
Austria
Orders LinkContactUsLinkComponent
Mini cable lockouts

Mini cable lockouts

Supplier:   Brady
Mini cable lockouts
Supplier:  Brady
Certificates
Mini cable lockouts
Mini cable lockouts

2 Options Available Below

Product Details & Documents

These mini cable lockouts are perfect for use on disconnect switches, small gate valves, etc.

  • Available with 2,44 m nylon or sheathed steel cable
  • Sheathed steel cable provides extra strength for a tight fit on valves and levers
  • 6 lock holes accommodate most padlock shackles and hasps
  • Push-button self-winding mechanism automatically cinches cable tight

Max. shackle diameter: 7,00 mm

Min. shackle clearance: 20,00 mm

Max. wire/cable diameter: 1,60 mm

Product Family Options

Product Information

  • Type
    Colour
    Availability
    Price
  • With steel cable (2,44 m)
    Red
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 85,40 €
    85,40 €
    1 * 1 ST
       85,40 €

  • With nylon cable (2,44 m)
    Red
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,20 €
    89,20 €
    1 * 1 ST
       89,20 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.