Orders LinkContactUsLinkComponent
Single-point sampler
Supplier:   Sampling Systems
Single-point sampler
Supplier:  Sampling Systems
Certificates
Single-point sampler
Single-point sampler
2 Options Available Below
Product Details & Documents
Single-point sampler for ice, wax, and similar materials.
Product Family Options
Product Information
  • Material
    Length
    Ø
    Volume
    Availability
    Price
  • SAMP6000A-550
    Ice sampler
    Ice sampler
    Stainless steel 316L
    550 mm
    40 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 553,00 €
    553,00 €
    1 * 1 ST
       553,00 €
  • SAMP6000A-1050
    Ice sampler
    Ice sampler
    Stainless steel 316L
    1050 mm
    40 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 603,00 €
    603,00 €
    1 * 1 ST
       603,00 €