About this item
Semi-micro balances that have 5" colour touch screen display with advanced applications and large databases for professional weighing. The user has easy, intuitive access to balance functions, via interactive, icon-driven menus ensuring efficient and productive weighing. Balances can be used by up to 10 different users, each with their own customisable settings and password protection. Ideal for laboratories requiring GLP compliance. All models have a stainless steel top pan.
- Internal and external calibration (optional factory calibration certificate)
- Multiple languages (English, German, French, Italian, Spanish or Portuguese)
- Check weighing, statistics, dynamic weighing, formulation and advanced piece counting function with statistics report
- Advanced density function, with density index calculation for quality assessment of metals
- Special textile function to measure fibres/filaments (cotton, linen, wool, nylon and polyester) with statistics report
- Serial output for connection to printer, USB port for database import or export and test results
User can choose three options for models with internal calibration (denoted by 'i' at end of the model's name):
Balance will calibrate using the internal weight by pressing CAL button
Balance will automatically recalibrate when ambient temperature changes and/or after a certain time has passed (Incal+)
The user can choose to calibrate the balance with an external weight
Related Information
Avantor Cell Culture Solutions.pdf VWR® for Weighing
Produktliteratur
Manual Touchscreen Balances