ContactUsLinkComponent
Multicoated microplates
Multicoated microplates
Catalog # CORN3829
Supplier:  Corning
undefined
Multicoated microplates
Catalog # CORN3829
Supplier:  Corning
Specifications
  • Recommended working volume:
    10 μl
  • Sterile:
    -
  • Colour:
    Black/clear
  • Description:
    Flat bottom, low base, with lid
  • Seq:
    02
  • Pk:
    10
  • Well volume:
    16,5 µl
Specifications
About this item
The Corning® multicoated microplate provides users access to six different surface treatments on a single plate.
This microplate is useful when determining the correct surface for assay requirements. Single surface microplates can then be used for the full screen or experiment. These multicoated plates are currently available in 96-well, 384-well, and 1536-well formats in black with clear bottom.