These plain white non sterile microfiltration membranes are used to remove particles and microorganisms from various organic solvents and aqueous solutions.
- Particle retention and collection
- Sterile filtration of solutions
- Particulate analysis
- Air monitoring applications
- Microbiological analysis of potable and waste waters
Cellulose acetate: Low protein-binding, well suited for sterile filtration and clarification of aqueous solutions in the pH range 4 - 8, nutrient media, buffers, and sera. Also compatible with alcohol and oil. These hydrophilic membranes offer high and consistent flow rates. Maximum thermal stability is 180 °C.
Cellulose nitrate: Set standard in both analytical and microbiological filtration. Available in a wide range of pore size and is well suited for microbial analysis in water, food and beverage application in the pH range 4 - 8. Maximum thermal stability is 130 °C. These membranes are available in a 47 and 50 mm ready-to-use standard size and it supports optimum colony growth.
Regenerated cellulose: Hydrophilic, pH range 3 to 12 and resistant to solvents. Typically used for HPLC sample preparation. Maximum thermal stability is 200 °C.
Nylon (Polyamide): Hydrophilic and widely used in both aqueous in the pH range 3 to 14 and organic solvent filtration applications. Well suited for clarification of buffers and nutrient media with a low level of extractables. Maximum thermal stability is 100 °C.
PTFE: Permanently hydrophobic filter material, very suitable for air and gas particulate filtration. Extremely resistant to aggressive solvents and acids in the pH range 1 to 14. Maximum thermal stability is 200 °C. Must be pre-wetted with ethanol before the filtration of aqueous media.
- Catalog No:
- 516-0791
- 516-0790
- 516-0787
- 516-0798
- 516-0786
- 514-0793
- 516-0789
- 516-0785
- 516-0788
- 516-0794
- 516-0793
- 516-0796
- 516-0795
- 516-0784
- 516-0792
- 516-0782
- 516-0783
- 516-0797
- 514-0732
- 514-0792
- 514-0794
- Pore size:
- 8,00 µm
- 0,45 µm
- 0,45 µm
- 5,00 µm
- 1,20 µm
- 0,45 µm
- 0,45 µm
- 0,20 µm
- 0,20 µm
- 0,80 µm
- 1,20 µm
- 0,45 µm
- 0,65 µm
- 0,45 µm
- 3,00 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,45 µm
- 0,80 µm
- Water flow rate:
- 750 ml/min/cm²
- >35 ml/min/cm²
- 0,2 ml/min/cm²
- 570 ml/min/cm²
- 1,6 ml/min/cm²
- 20 ml/min/cm²
- 30 ml/min/cm²
- 24 ml/min/cm²
- 0,1 ml/min/cm²
- 200 ml/min/cm²
- 320 ml/min/cm²
- 70 ml/min/cm²
- 130 ml/min/cm²
- 65 ml/min/cm²
- 430 ml/min/cm²
- 65 ml/min/cm²
- 65 ml/min/cm²
- 70 ml/min/cm²
- 70 ml/min/cm²
- 69 ml/min/cm²
- 200 ml/min/cm²
- Ø:
- 47 mm
- 47 mm
- 47 mm
- 47 mm
- 47 mm
- 25 mm
- 47 mm
- 47 mm
- 47 mm
- 47 mm
- 47 mm
- 25 mm
- 47 mm
- 50 mm
- 90 mm
- 25 mm
- 47 mm
- 50 mm
- 47 mm
- 90 mm
- 50 mm