ContactUsLinkComponent
Red long-term cell tracer, CYTO-ID®
Red long-term cell tracer, CYTO-ID®
  ENZOENZ51037K025
 :  
Red long-term cell tracer, CYTO-ID®
  ENZOENZ51037K025
 :  
  • Pk:
    1 KIT
  • Description:
    Red long-term cell tracer
  • Seq:
    02
CYTO-ID® is a live cell fluorescent labelling used for a long term cell viability, cytotoxicity, cell adhesion, cell migration, and cell-cell fusion studies.
  • Dual labeling with CELLESTIAL® fluorescent probes
  • Minimal transfer of fluorescence from dye-labeled to unlabelled cells
Uses proprietary non covalent cell labeling technology to stably incorporate a red fluorescent dye containing hydrophobic aliphatic chains into the cell membrane's lipid bilayer.
 : Contains a red tracer dye (50 µl), 4X labeling buffer (12,5 ml), and 10X HBSS (25 ml).
 : Kit contains CYTO-ID® red tracer dye, 50 µl, 4X labeling buffer, 12,5 ml and 10X HBSS, 25 ml.