ContactUsLinkComponent
Open end wrenches
Open end wrenches
Catalog # REST20388
Supplier:  Restek
undefined
Open end wrenches
Catalog # REST20388
Supplier:  Restek
Specifications
  • Description:
    Open end wrench, 4,8 - 6,4 mm
  • Pk:
    2
Specifications
About this item
High-quality miniature wrench to be used with MXT® low-dead-volume connectors.