Capillary columns, Rtx®-VRX (fused silica)
Supplier: Restek
Certificates
About this item
Proprietary Crossbond® phase application-specific columns for volatile organic pollutants.
- Suitable for U.S. EPA Method 8021 compounds
- Stable to 260 °C
Stationary phase and optimised column dimensions provide low bleed, excellent resolution, and fast analysis times for volatile compounds.
Frequently Bought Together