ContactUsLinkComponent
Universal cylinder wrench
Universal cylinder wrench
Catalog # REST21322
Supplier:  Restek
undefined
Universal cylinder wrench
Catalog # REST21322
Supplier:  Restek
Specifications
  • Description:
    Universal cylinder wrench
  • Pk:
    1
Specifications
About this item
This versatile wrench is used for tightening gauges and gas regulator CGA fittings to cylinder outlets and pipe thread connections.