ContactUsLinkComponent
Qualitative filter papers, grade 595, Whatman™
Qualitative filter papers, grade 595, Whatman™
Certificates
undefined
Qualitative filter papers, grade 595, Whatman™
Specifications
  • Catalog No:
  • 512-3618
  • 515-5513
  • 515-4505
  • 512-3619
  • Grade:
  • 595
  • 595
  • 595
  • 595
  • Weight:
  • 68 g/m²
  • 68 g/m²
  • 68 g/m²
  • 68 g/m²
  • Thickness:
  • 150 µm
  • 150 µm
  • 150 µm
  • 150 µm
  • Particle retention:
  • 7 µm
  • 7 µm
  • 7 µm
  • 7 µm
  • Filtration speed:
  • 80 s (Herzberg)
  • 80 s (Herzberg)
  • 80 s (Herzberg)
  • 80 s (Herzberg)