ContactUsLinkComponent
Qualitative filter papers, standard grades, grade 593 ¹/₂, Whatman™
Qualitative filter papers, standard grades, grade 593 ¹/₂, Whatman™
Certificates
undefined
Qualitative filter papers, standard grades, grade 593 ¹/₂, Whatman™
Specifications
  • Catalog No:
  • 515-5607
  • 515-5606
  • Grade:
  • 593 ½
  • 593 ½
  • Weight:
  • 85 g/m²
  • 85 g/m²
  • Thickness:
  • 170 µm
  • 170 µm
  • Particle retention:
  • 5 µm
  • 5 µm
  • Filtration speed:
  • 450 s (Herzberg)
  • 450 s (Herzberg)