Syringe filters, nylon
Catalog # BINZ6012N13002
Supplier: Ahlstrom-Munksjö
Specifications
- Ø:13 mm
- Membrane:Nylon
- Pore size:0,20 µm
- Sterility:Non sterile
- Type:Hydrophilic
- Environmentally Preferable:
- Pk:100
Specifications
About this item
This syringe filter with hydrophilic polyamide membrane is used for sterilisation and clarification of biological fluids, buffer solutions and tissue culture medias. Also suitable for filtration of aqueous and organic solutions in HPLC.
- Good mechanical strength and strong absorption
- Resistance against most of organic solvents and most of alkaline liquids
- Chemical compatibility: pH ranges from 3 to 14
- Filtration and clarification of solvents