Capillary columns, Rtx®-200MS (fused silica)
About this item
Rtx-200 columns have accomplished many difficult separations. The trifluoropropylmethyl polysiloxane stationary phase has a unique selectivity that changes elution orders and resolves compounds that phenyl, cyano, or carbowax phases can not. These columns offer exceptional thermal stability, low bleed, and superior inertness - even for active compounds such as phenols, and with sensitive detectors such as ECDs, NPDs, and MSDs.
- Equivalent to USP G6 phase
General purpose columns for solvents, Freon fluorocarbons, alcohols, ketones, silanes, glycols. Excellent confirmation column with an Rtx-5 column for phenols, nitrosamines, organochlorine pesticides, chlorinated hydrocarbons, and chlorophenoxy herbicides.
26 Options Available Below
Product Details & Documents
Rtx-200 columns have accomplished many difficult separations. The trifluoropropylmethyl polysiloxane stationary phase has a unique selectivity that changes elution orders and resolves compounds that phenyl, cyano, or carbowax phases can not. These columns offer exceptional thermal stability, low bleed, and superior inertness - even for active compounds such as phenols, and with sensitive detectors such as ECDs, NPDs, and MSDs.
- Equivalent to USP G6 phase
General purpose columns for solvents, Freon fluorocarbons, alcohols, ketones, silanes, glycols. Excellent confirmation column with an Rtx-5 column for phenols, nitrosamines, organochlorine pesticides, chlorinated hydrocarbons, and chlorophenoxy herbicides.
Product Family Options
Product Information
- Column FormatPhaseFilm thicknessØ int.LengthAvailabilityPrice
- REST15605Rtx-200MS cap column
- REST15620Rtx-200MS cap column
- JJBEI15635Rtx-200MS cap columnSpecifications:MarketsourceMarketsource
- REST15650Rtx-200MS cap column
- REST15608Rtx-200MS cap column
- REST15623Rtx-200MS cap column
- REST15638Rtx-200MS cap column
- JJBEI15653Rtx-200MS cap columnSpecifications:MarketsourceMarketsource
- REST15606Rtx-200MS cap column
- REST15621Rtx-200MS cap column
- REST15636Rtx-200MS cap column
- REST15651Rtx-200MS cap column
- REST15609Rtx-200MS cap column
- REST15624-600Rtx-200MS cap column
- REST15624-6850Rtx-200MS cap column with 127 mm HP6850 cageRtx-200MS cap column with 127 mm HP6850 cageCapillaryRtx®-200MS0,25 µm0,32 mm30 mLog in for availability1 * 1 ST
- REST15624Rtx-200MS cap column
- REST15639Rtx-200MS cap column
- REST15654Rtx-200MS cap column
- REST15637Rtx-200MS cap column
- REST15652-282Rtx-200MS cap column with 127 mm star cage - no guarantee for breakageRtx-200MS cap column with 127 mm star cage - no guarantee for breakageMicroboreRtx®-200MS1,00 µm0,53 mm15 mLog in for availability1 * 1 ST
- REST15652Rtx-200MS cap column
- REST15667Rtx-200MS cap column
- REST15640Rtx-200MS cap column
- REST15655-6850Rtx-200MS cap column with 127 mm HP6850 cageRtx-200MS cap column with 127 mm HP6850 cageMicroboreRtx®-200MS1,00 µm0,53 mm30 mLog in for availability1 * 1 ST
- REST15655Rtx-200MS cap column
- REST15670Rtx-200MS cap column